Learn More
Eppendorf™ Rotor for Centrifuge 5810 / 5810 R
Shop All Eppendorf Products
Click to view available options
Capacity:
2 x 4 MTP, 2 x 1 DWP
4 x 750 mL
Max. Speed:
3,900 rpm
4,500 rpm
Max. RCF:
3,214 x g
3,486 x g
Includes:
750 mL Round Buckets (x4)
Buckets (x2), Aerosol-tight Caps (x2), Plate Carriers (x2)
Plate Buckets (x4)
Without Buckets
Unit Size:
Each
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
Specifications
Specifications
| Description | A-2-DWP-AT - Includes buckets (x2), aerosol-tight caps (x2), plate carriers (x2) |
| Type | Rotor |
| Capacity | 2 x 4 MTP, 2 x 1 DWP |
| Max. RCF | 3,486 x g |
| Max. Speed | 4,500 rpm |
| For Use With (Equipment) | Centrifuge 5810 / 5810 R |
| Includes | Buckets (x2), Aerosol-tight Caps (x2), Plate Carriers (x2) |