missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H2R Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS1176
This item is not returnable.
View return policy
Description
Histamine H2R Polyclonal antibody specifically detects Histamine H2R in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Histamine H2R | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 18 μg/mL | |
| gastric receptor 1, Gastric receptor I, H2R, HH2R, histamine H2 receptor, histamine receptor H2 | |
| Synthetic 16 amino acid peptide from 3rd extracellular domain of human Histamine H2R. | |
| 0.05 mL | |
| GPCR, Stem Cell Markers | |
| 3274 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction