missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Annexin A2 Protein

Product Code. 18755598
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18755598

Brand: R&D Systems™ 9409AN050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium-dependent phospholipid-binding proteins.

The Recombinant Human Annexin A2 Protein is derived from E. coli.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07355-1
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS and DTT
Gene ID (Entrez) 302
Molecular Weight (g/mol) 39 kDa
Quantity 50 μg
Source E. coli-derived human Annexin A2 protein Ser2-Asp339,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias ANX2, ANX2L4, ANX2L4LPC2, ANXA2, CAL1H
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.