missing translation for 'onlineSavingsMsg'
Learn More

(S)-Mephenytoin, TRC

Product Code. 17933125
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933125 50 mg 50mg
30011138 100 mg 100mg
17982925 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933125 Supplier Toronto Research Chemicals Supplier No. TRCM22500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70989-04-7
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.25
Synonym 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-, (5S)-, 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-, (S)-, (5S)-5-Ethyl-3-methyl-5-phenyl-2,4-imidazolidinedione, (+)-5-Ethyl-3-methyl-5-phenylhydantoin, (+)-Mephenytoin, (+)-Mesantoin, (+)-S-Mephenytoin, (S)-5-Ethyl-3-methyl-5-phenylimidazolidine-2,4-dione, (S)-Mephenytoin, d-Mephenytoin
IUPAC Name (5S)-5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
SMILES CC[C@]1(NC(=O)N(C)C1=O)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)/t12-/m0/s1
Formula Weight 218.1055
Percent Purity >95%
Chemical Name or Material (S)-Mephenytoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.