missing translation for 'onlineSavingsMsg'
Learn More

US EPA ICP-MS ICS A Mix 1: Al, Ca, Fe, K, Mg, Na, P, S @ 500; C @ 1000; Cl @ 5000 μg/mL in 5% HNO3, 1.5% HCl, VHG™

Product Code. 17917839
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
missing translation for 'unitSize'
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 17917839

missing translation for 'mfr': VHG Labs VHGLICSA1Z500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

US EPA ICP-MS ICS A Mix 1: Al, Ca, Fe, K, Mg, Na, P, S @ 500; C @ 1000; Cl @ 5000 μg/mL in 5% HNO3, 1.5% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Calcium/Iron/Magnesium/Phosphorus/Potassium/Sodium/Sulfur/Carbon/Chlorine
CAS 7429-90-5, 7440-70-2, 7439-89-6, 7439-95-4, 7723-14-0, 9-7-7440
Analyte or Component Names Aluminium/Calcium/Iron/Magnesium/Phosphorus/Potassium/Sodium/Sulfur/Carbon/Chlorine
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3/HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Aluminium 500 μg/mL/Calcium 500 μg/mL/Iron 500 μg/mL/Magnesium 500 μg/mL/Phosphorus 500 μg/mL/Potassium 500 μg/mL/Sodium 500 μg/mL/Sulfur 500 μg/mL/Carbon 1000 μg/mL/Chlorine 5000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.