missing translation for 'onlineSavingsMsg'
Learn More

Chem-Lab 500ML POTASSIUM THIOCYANATE 5% SOLUTION

Brand:  Chem-Lab CL12.1104.0500


500ML POTASSIUM THIOCYANATE 5% SOLUTION

Product Code. 13466108

  • 21.75 EUR / 500mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium thiocyanate 5% solution

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

Supplemental information
  • MIXTURE LIST-Contains : Potassium thiocyanate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.