missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkalis & Alkaline Earths Standard: Ba, Be, Ca, Cs, K, Li, Mg, Na, Rb, Sr @ 100 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Alkalis & Alkaline Earths Standard: Ba, Be, Ca, Cs, K, Li, Mg, Na, Rb, Sr @ 100 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Calcium/Strontium/Lithium/Barium/Beryllium/Magnesium/Potassium/Rubidium/Sodium/Caesium |
| CAS | 7440-70-2, 7440-24-6, 7439-93-2, 7440-39-3, 7440-41-7, 7439-95-4 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Calcium/Strontium/Lithium/Barium/Beryllium/Magnesium/Potassium/Rubidium/Sodium/Caesium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Calcium 99.98 μg/mL/Strontium 99.98 μg/mL/Lithium 99.99 μg/mL/Barium 100 μg/mL/Beryllium 100 μg/mL/Magnesium 100 μg/mL/Potassium 100 μg/mL/Rubidium 100 μg/mL/Sodium 100 μg/mL/Caesium 100.1 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction