missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxeladin Hydrogen Citrate, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Oxeladin Hydrogen Citrate
Specifications
Specifications
| Chemical Name or Material | Oxeladin hydrogen citrate |
| CAS | 52432-72-1 |
| Analyte or Component Names | Oxeladin hydrogen citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H33 N O3 . C6 H8 O7 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Oxeladin Hydrogen Citrate, 2-Ethyl-2-phenylbutyric acid 2-(2-diethylaminoethoxy)ethyl ester citrate, Oxeladin citrate, Oxeladine citrate, Paxeladine, Pectamol, Petcamon, Silopentol, alpha,alpha-Diethylbenzeneacetic acid 2-[2-(diethylamino)ethoxy]ethyl ester 2-hydroxy-1,2,3-propanetricarboxylate (1:1) |
| InChI Formula | InChI=1S/C20H33NO3.C6H8O7/c1-5-20(6-2,18-12-10-9-11-13-18)19(22)24-17-16-23-15-14-21(7-3)8-4;7-3(8)1-6(13,5(11)12)2-4(9)10/h9-13H,5-8,14-17H2,1-4H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CCN(CC)CCOCCOC(=O)C(CC)(CC)c1ccccc1.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 2-[2-(diethylamino)ethoxy]ethyl 2-ethyl-2-phenylbutanoate;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction