Learn More
Labcon™ MetalFree™ 50 mL Polypropylene Centrifuge Tubes
Click to view available options
Packaging:
25 Tubes/Low Waste Fiber Rack, 20 Racks/Cs.
25 Tubes/Rack, 20 Rcks/Cs
50 Tubes/Bag, 10 Bags/Cs.
Unit Size:
Pack of 500
Description
- Conical centrifuge tube
- With attached flat screw caps
- Certified free of metals to 1 part per billion of 43 common metals and elements
- Caps are pure natural polyethylene
- Patented anti-smear writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Polypropylene USP Class VI resin
- Centrifuging to 17,000 x G
- Freezable to -80°C
- Autoclavable
- Capacity: 50 mL
- Natural colorless caps
- Made in USA
- Elements tested: Aluminium, Antimony, Arsenic, Barium,, Beryllium, Bismuth, Cadnium, Caesium, Calcium, Chromium, Cobalt, Copper, Gallium, Germanium, Gold, Iron, Lead , Lithium, Magnesium, Manganese, Mercury, Molebdenum, Nickel, Niobium, Phosphorus, Potassium, Rhenium, Rubidium, Selenium, Silicon, Silver, Sodium, Strontium, Tantalum, Thallium, Tin, Titanium, Tungsten, Vanadium, Yttrium, Zinc, Zirconium
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015, FDA CFR21 GMP |
| Closure Material | Polyethylene |
| Closure Color | Natural |
| Capacity (Metric) | 50 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| For Use With | Centrifuges, rotators, shakers, ICP-MS |
| No. per Pack | 50/Pk. |
| Packaging | 50 Tubes/Bag, 10 Bags/Cs. |
| Sterility | Non-sterile |
| Show More |