Learn More
Hamilton™ 50 μL 1800 Series Gas Tight Syringe
Product Code. 17854232
Change view
Click to view available options
Needle Gauge:
22s G
No Needle Included
Needle Point Style:
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Removable
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Needle Point Style Connection Type unitSize
17854232 22s G Sharp, Bevel, Curved, Non-coring Cemented Each
17864282 22s G Sharp, Bevel, Curved, Non-coring Removable Each
17884212 No Needle Included No Needle Included Removable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17854232 Supplier Hamilton™ Supplier No. CAL763801
Only null left
Add to basket
Add to basket
The 1800 Series Gas Tight syringes are the reinforced plunger version of the 1700 Series.
  • Attached to the flange of the syringe is a blue syringe holder, which provides an area to hold the syringe that prevents heat transfer, dispense inaccuracies, and plunger blow outs.
  • Provides additional support to small-volume plungers that are prone to bending during injection.
  • This series of syringes are ideal for dispensing volumes up to 50 μL.
Specifications
Autoclavable Not Autoclavable
Material PTFE
Needle Gauge 22s G
Diameter (English) Inner 0.029 in.
Diameter (English) Inner Needle 0.0059 in.
Diameter (Metric) Inner Needle 0.15 mm
Diameter (English) Outer Needle 0.0283 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 0.729 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
Length (English) Needle 2 in.
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 1805 N
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 50 μL
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Cemented