missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5% (w/w) Devolatilized Diesel Fuel in 75 cSt Hydrocarbon Oil, VHG™
Description
5% (w/w) Devolatilized Diesel Fuel in 75 cSt Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Devolatilized Diesel Fuel |
| Methods | IR/Fuel Sniffer/Flash Point Analyzer |
| Analyte or Component Names | Devolatilized Diesel Fuel |
| Shipping Condition | Room Temperature |
| Concentration | 5% |
| Synonym | Devolatilized Diesel Fuel |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?