Learn More
5-pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine, Thermo Scientific™
Product Code. 10577780
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10577780 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10577780 Supplier Thermo Scientific Maybridge Supplier No. MAY00308DA
Request Bulk or Custom Format
Specifications
CAS 71125-45-6
Packaging Amber Glass Bottle
Molecular Formula C6H10N4S
Quantity 1 g
Synonym 5-pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine, tos-bb-1082, 2-amino-5-pyrrolidino-1,3,4-thiadiazole, 5-pyrrolidinyl-1,3,4-thiadiazole-2-ylamine, 5-1-pyrrolidinyl-1,3,4-thiadiazol-2-amine, 1,3,4-thiadiazol-2-amine, 5-1-pyrrolidinyl, 5-pyrrolidin-1-yl-1,3,4 thiadiazol-2-ylamine
InChI Key UWZJRWBCNZFSMS-UHFFFAOYSA-N
SMILES C1CCN(C1)C2=NN=C(S2)N
IUPAC Name 5-pyrrolidin-1-yl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol) 170.234
PubChem CID 1500014
Formula Weight 170.23
Percent Purity 90%
Chemical Name or Material 5-pyrrolidin-1-yl-1, 3, 4-thiadiazol-2-amine