Learn More
Applied Biosystems™ 5' Phosphate Labeled Oligo
Shop All Applied Biosystems Products
Click to view available options
Quantity:
80,000 pmol
Unit Size:
80000pmol
Description
5' Phosphate labeled oligo
Specifications
Specifications
| Product Type | Oligonucleotides |
| Length (Metric) | 1.524 m |
| Content And Storage | Store in Freezer (-5°C to -30°C). |
| Label or Dye | 6-FAM, HEX, NED, PET, TET, VIC |
| Primer | 5' Phosphate Labeled |
| Quantity | 80,000 pmol |