missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo-5-(2-thienyl)valeric acid, 97%, Thermo Scientific™

Product Code. 11368076
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11368076

Brand: Thermo Scientific Alfa Aesar L13362.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22971-62-6
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
MDL Number MFCD00151848
InChI Key GSQMRVFOHNXKOR-UHFFFAOYSA-N
Synonym 5-oxo-5-2-thienyl valeric acid, 5-oxo-5-thiophen-2-yl pentanoic acid, 4-2-thienoyl butyric acid, 5-oxo-5-thien-2-ylpentanoic acid, 5-oxo-5-2-thienyl pentanoic acid, 2-thiophenepentanoicacid, d-oxo, acmc-1cj8s, 4-2-thenoyl butyric acid, 5-2-thienoyl butyric acid, 5-thien-2-yl-5-oxopentanoic acid
PubChem CID 255155
IUPAC Name 5-oxo-5-thiophen-2-ylpentanoic acid
SMILES C1=CSC(=C1)C(=O)CCCC(=O)O

Specifications

Melting Point 94°C to 97°C
Quantity 5 g
Beilstein 141775
Formula Weight 198.24
Percent Purity 97%
Chemical Name or Material 5-Oxo-5-(2-thienyl)valeric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.