Learn More
5-O-Demethyl-28-Oxo-Avermectin A1A, TRC
Specifications
Specifications
| Chemical Name or Material | 5-O-Demethyl-28-Oxo-Avermectin A1A |
| CAS | 102190-68-1 |
| Molecular Formula | C48 H70 O15 |
| Quantity | 5 mg |
| Synonym | Spiro[11,15-Methano-2H,13H,17H-Furo[4,3,2-Pq] |
| InChI Formula | InChI=1S/C48H70O15/c1-11-24(2)42-27(5)17-18-47(63-42)23-32-20-31(62-47)16-15-26(4)41(59-38-22-36(55-10)43(30(8)57-38)60-37-21-35(54-9)40(50)29(7)56-37)25(3)13-12-14-33-45(51)61-44-39(49)28(6)19-34(46(52)58-32)48(33,44)53/h12-15,17-19,24-25,27,29-32,34-44,49-50,53H,11,16,20-23H2,1-10H3/b13-12+,26-15+,33-14+/t24-,25-,27-,29-,30-,31+,32-,34-,35-,36-,37-,38-,39+,40-,41-,42+,43-,44+,47+,48+/m0/s1 |
| SMILES | CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/C(=O)O[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C |
| IUPAC Name | (1'R,2R,3S,4'S,6S,8'R,10'E,12'S,13'S,14'E,16'Z,20'R,21'R,24'S)-2-[(2S)-butan-2-yl]-21',24'-dihydroxy-12'-[(2R,4S,5S,6S)-5-[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy-4-methoxy-6-methyloxan-2-yl]oxy-3,11',13',22'-tetramethylspiro[2,3-dihydropyran-6,6'-3,7,19-trioxatetracyclo[15.6.1.14,8.020,24]pentacosa-10,14,16,22-tetraene]-2',18'-dione |
| Molecular Weight (g/mol) | 887.06 |
| Formula Weight | 886.471 |