missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanol, TRC

Product Code. 30478592
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478592 10 g 10g
30474034 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30478592 Supplier Toronto Research Chemicals Supplier No. TRCM31208010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-98-5
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.2
Synonym 5-Methyl-1-hexanol
IUPAC Name 5-methylhexan-1-ol
SMILES CC(C)CCCCO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H16O/c1-7(2)5-3-4-6-8/h7-8H,3-6H2,1-2H3
Formula Weight 116.1201
Chemical Name or Material 5-Methylhexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.