Learn More
5-Methyl-2-hexanone, 99%
Product Code. 11308536
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
Product Code. 11308536
Brand: Thermo Scientific Alfa Aesar L17584.0F
Request Bulk or Custom Format
CAS: 110-12-3 | C7H14O | 114.188 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water. (5.4 g/L)
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C
Specifications
Melting Point -74°C
Density 0.812
Boiling Point 144°C to 145°C
Flash Point 41°C (105°F)
Refractive Index 1.4075
Quantity 2500 mL
UN Number UN2302
Beilstein 506163
Solubility Information Soluble in water. (5.4g/L)
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hexanone