missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxypyrazinecarboxamide, TRC

Product Code. 30003275
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003275 250 mg 250mg
30004873 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003275 Supplier Toronto Research Chemicals Supplier No. TRCH952745250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13924-96-4
Molecular Formula C5 H5 N3 O2
Molecular Weight (g/mol) 139.11
Synonym 2-Carboxamido-5-hydroxypyrazine, 5-Hydroxypyrazinecarboxamide, 4,5-Dihydro-5-oxopyrazinecarboxamide
IUPAC Name 6-oxo-1H-pyrazine-3-carboxamide
SMILES NC(=O)C1=CNC(=O)C=N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C5H5N3O2/c6-5(10)3-1-8-4(9)2-7-3/h1-2H,(H2,6,10)(H,8,9)
Formula Weight 139.0382
Chemical Name or Material 5-Hydroxypyrazinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.