missing translation for 'onlineSavingsMsg'
Learn More

(5-Chloro-2-nitrophenyl)(phenyl)carbamic Acid tert-Butyl Ester, TRC

Product Code. 30359539
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359539 500 mg 500mg
30349789 250 mg -
30358442 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359539 Supplier Toronto Research Chemicals Supplier No. TRCC368545500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C17H17ClN2O4
Molecular Weight (g/mol) 348.78
IUPAC Name tert-butyl (5-chloro-2-nitrophenyl)(phenyl)carbamate
SMILES O=C(OC(C)(C)C)N(C1=CC(Cl)=CC=C1[N+]([O-])=O)C2=CC=CC=C2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H17ClN2O4/c1-17(2,3)24-16(21)19(13-7-5-4-6-8-13)15-11-12(18)9-10-14(15)20(22)23/h4-11H,1-3H3
Formula Weight 348.09
Chemical Name or Material (5-Chloro-2-nitrophenyl)(phenyl)carbamic Acid tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.