missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxy-4-methylaminobenzoic Acid, TRC

Product Code. 30359687
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30359687 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359687 Supplier Toronto Research Chemicals Supplier No. TRCC36865025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-methoxy-4-methylaminobenzoic Acid
CAS 61694-98-2
Molecular Formula C9 H10 Cl N O3
Quantity 25 mg
InChI Formula InChI=1S/C9H10ClNO3/c1-11-7-4-8(14-2)5(9(12)13)3-6(7)10/h3-4,11H,1-2H3,(H,12,13)
SMILES CNc1cc(OC)c(cc1Cl)C(=O)O
IUPAC Name 5-chloro-2-methoxy-4-(methylamino)benzoic acid
Molecular Weight (g/mol) 215.63
Formula Weight 215.0349
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.