Learn More
5-Chloro-2-fluoroaniline, 97%
Product Code. 11460007
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11460007 10 g 10g
11470007 50 g 50g
2 options
Product Code. 11460007
Brand: Thermo Scientific Alfa Aesar B20951.09
Request Bulk or Custom Format
CAS: 2106-05-0 | C6H5ClFN | 145.561 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2106-05-0
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00069416
InChI Key JCYROOANFKVAIB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine
PubChem CID 75015
IUPAC Name 5-chloro-2-fluoroaniline
SMILES C1=CC(=C(C=C1Cl)N)F
Specifications
Density 1.3
Boiling Point 102°C to 105°C (20 mmHg)
Quantity 10 g
Odor Characteristic
Refractive Index 1.56
UN Number UN2810
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluoroaniline