missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide, 98%, Thermo Scientific™

Product Code. 11431818
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431818 5 g 5g
11421818 1 g 1g
11441818 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431818 Supplier Thermo Scientific Alfa Aesar Supplier No. A14487.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to heat, light and moisture. Handle and store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167683-86-5
Molecular Formula C4H7Br2N3
Molecular Weight (g/mol) 256.929
MDL Number MFCD00053023
InChI Key UGCOULAJNKWOPX-UHFFFAOYSA-N
Synonym 5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide
PubChem CID 2778427
IUPAC Name 4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide
SMILES CC1=C(C(=NN1)N)Br.Br

Specifications

Melting Point 178°C to 181°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 256.94
Percent Purity 98%
Chemical Name or Material 5-Amino-4-bromo-3-methyl-1H-pyrazole hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.