Learn More
5-Acetamidonaphthalene-1-sulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Acetamidonaphthalene-1-sulfonyl Chloride |
| CAS | 52218-37-8 |
| Molecular Formula | C12 H10 Cl N O3 S |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C12H10ClNO3S/c1-8(15)14-11-6-2-5-10-9(11)4-3-7-12(10)18(13,16)17/h2-7H,1H3,(H,14,15) |
| SMILES | CC(=O)Nc1cccc2c(cccc12)S(=O)(=O)Cl |
| IUPAC Name | 5-acetamidonaphthalene-1-sulfonyl chloride |
| Molecular Weight (g/mol) | 283.73 |
| Formula Weight | 283.007 |