missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichlorokynurenic acid

Product Code. 15423779
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 15423779

Brand: Thermo Scientific Alfa Aesar J61138.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 190908-40-8 | C10H7Cl2NO4 | 276.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190908-40-8
Molecular Formula C10H7Cl2NO4
Molecular Weight (g/mol) 276.069
MDL Number MFCD11045951
InChI Key READYYZXDSXRQJ-UHFFFAOYSA-N
Synonym 5,7-dichlorokynurenic acid monohydrate, dcka hydrate, c10h5cl2no3 hydrate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid monohydrate, 5,7-dichlorokynurenic acid monohydrate, solid hplc, 2-quinolinecarboxylic acid, 5,7-dichloro-4-hydroxy-, monohydrate, 2-quinolinecarboxylicacid, 5,7-dichloro-4-hydroxy-, hydrate 1:1, 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid-water 1/1
PubChem CID 23723057
IUPAC Name 5,7-dichloro-4-oxo-1H-quinoline-2-carboxylic acid;hydrate
SMILES C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)O)Cl.O

Specifications

Color Yellow
Quantity 100 mg
Solubility Information Soluble in dilute sodium hydroxide
Formula Weight 258.06
Physical Form Solid
Chemical Name or Material 5,7-Dichlorokynurenic acid monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.