missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichlorokynurenic acid

Product Code. p-7042035
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413779 10 mg 10mg
15433779 50 mg 50mg
15423779 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15413779 Supplier Thermo Scientific Alfa Aesar Supplier No. J61138.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 190908-40-8 | C10H7Cl2NO4 | 276.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190908-40-8
Molecular Formula C10H7Cl2NO4
Molecular Weight (g/mol) 276.069
MDL Number MFCD11045951
InChI Key READYYZXDSXRQJ-UHFFFAOYSA-N
Synonym 5,7-dichlorokynurenic acid monohydrate, dcka hydrate, c10h5cl2no3 hydrate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid monohydrate, 5,7-dichlorokynurenic acid monohydrate, solid hplc, 2-quinolinecarboxylic acid, 5,7-dichloro-4-hydroxy-, monohydrate, 2-quinolinecarboxylicacid, 5,7-dichloro-4-hydroxy-, hydrate 1:1, 5,7-dichloro-4-oxo-1,4-dihydroquinoline-2-carboxylic acid-water 1/1
PubChem CID 23723057
IUPAC Name 5,7-dichloro-4-oxo-1H-quinoline-2-carboxylic acid;hydrate
SMILES C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)O)Cl.O

Specifications

Color Yellow
Quantity 10 mg
Solubility Information Soluble in dilute sodium hydroxide
Formula Weight 258.06
Physical Form Solid
Chemical Name or Material 5,7-Dichlorokynurenic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.