missing translation for 'onlineSavingsMsg'
Learn More

5,7,8,9-Tetrahydro-5-methyl-3-phenyl-pyrido[3',2':4,5]imidazo[1,2-a]pyrimidin-7-ol, TRC

Product Code. 30417912
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417912 1 mg 1mg
30423087 10 mg 10mg
30410394 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417912 Supplier Toronto Research Chemicals Supplier No. TRCT2931351MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,7,8,9-Tetrahydro-5-methyl-3-phenyl-pyrido[3',2':4,5]imidazo[1,2-a]pyrimidin-7-ol
Analyte or Component Names 5,7,8,9-Tetrahydro-5-methyl-3-phenyl-pyrido[3',2':4,5]imidazo[1,2-a]pyrimidin-7-ol
Shipping Condition Room Temperature
Molecular Formula C16 H16 N4 O
Synonym 5-Methyl-3-phenyl-5,7,8,9-tetrahydropyrido[3',2':4,5]imidazo[1,2-a]pyrimidin-7-ol
InChI Formula InChI=1S/C16H16N4O/c1-19-13-9-12(11-5-3-2-4-6-11)10-17-15(13)20-8-7-14(21)18-16(19)20/h2-6,9-10,14,21H,7-8H2,1H3
Solution Type Neat
SMILES CN1C2=NC(O)CCN2c3ncc(cc13)c4ccccc4
IUPAC Name 8-methyl-5-phenyl-1,3,8,10-tetrazatricyclo[7.4.0.02,7]trideca-2,4,6,9-tetraen-11-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 280.324
Quantity 1 mg
Formula Weight 280.132
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.