missing translation for 'onlineSavingsMsg'
Learn More

(4R)-tert-Butyldimethylsilyl 4-((3S,7R,8R,9S,10S,13R,14S,17R)-7-Hydroxy-10,13-dimethyl-3-((tetrahydro-2H-pyran-2-yl)oxy)hexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate, TRC

Product Code. 30343908
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343908 100 mg 100mg
30333217 5 mg 5mg
30342781 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30343908 Supplier Toronto Research Chemicals Supplier No. TRCB723435100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4R)-tert-Butyldimethylsilyl 4-((3S,7R,8R,9S,10S,13R,14S,17R)-7-Hydroxy-10,13-dimethyl-3-((tetrahydro-2H-pyran-2-yl)oxy)hexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate
Analyte or Component Names (4R)-tert-Butyldimethylsilyl 4-((3S,7R,8R,9S,10S,13R,14S,17R)-7-Hydroxy-10,13-dimethyl-3-((tetrahydro-2H-pyran-2-yl)oxy)hexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate
Shipping Condition Room Temperature
Molecular Formula C35 H62 O5 Si
InChI Formula InChI=1S/C35H62O5Si/c1-23(12-15-30(37)40-41(7,8)33(2,3)4)26-13-14-27-32-28(17-19-35(26,27)6)34(5)18-16-25(21-24(34)22-29(32)36)39-31-11-9-10-20-38-31/h23-29,31-32,36H,9-22H2,1-8H3/t23-,24?,25+,26-,27+,28+,29-,31?,32+,34+,35-/m1/s1
Solution Type Neat
SMILES C[C@H](CCC(=O)O[Si](C)(C)C(C)(C)C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)OC5CCCCO5
IUPAC Name [tert-butyl(dimethyl)silyl] (4R)-4-[(3S,7R,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-(oxan-2-yloxy)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 590.949
Quantity 100 mg
Formula Weight 590.437
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.