missing translation for 'onlineSavingsMsg'
Learn More

(4E, 8Z)-Sphingadienine-C18-1-phosphate, TRC

Product Code. 30427179
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427179 1 mg 1mg
30426333 10 mg 10mg
30420406 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427179 Supplier Toronto Research Chemicals Supplier No. TRCS6806551MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4E, 8Z)-Sphingadienine-C18-1-phosphate
Analyte or Component Names (4E, 8Z)-Sphingadienine-C18-1-phosphate
Shipping Condition Room Temperature
Molecular Formula C18H36NO5P
Synonym (2S,3R,4E,8Z)-2-Amino-3-hydroxyoctadeca-4,8-dien-1-yl Dihydrogen Phosphate, (E,Z)-D-erythro-2-Amino-4,8-octadecadiene-1,3-diol Dihydrogen Phosphate, [R-[R*,S*-(E,Z)]]-2-Amino-4,8-octadecadiene-1,3-diol Dihydrogen Phosphate
InChI Formula InChI=1S/C22H31BO3/c1-4-5-6-10-15-22(2,3)19-13-14-20(23(24)25)21(16-19)26-17-18-11-8-7-9-12-18/h7-9,11-14,16,24-25H,4-6,10,15,17H2,1-3H3
Solution Type Neat
SMILES CCCCCCC(C)(C)c1cc(OCc2ccccc2)c(B(O)O)cc1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 377.46
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.