Learn More
BRAND™ 48-Well Non-Skirted Microplates, BIO-CERT™ PCR QUALITY
Shop All Brand Products
Click to view available options
Color:
Clear
White
Unit Size:
Pack of 20
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Compact design is compatible with most standard thermal cyclers, ensuring convenient handling
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
Specifications
Specifications
| Barcode | No Barcode |
| Binding Property | Untreated |
| Certifications/Compliance | RNase, DNase, DNA-free |
| Format | Standard |
| Array | 6 x 8 |
| Autoclavable | Autoclavable |
| Color | Clear |
| Disposable | Single-use |
| Product Type | PCR 48-well plate |
| For Use With (Equipment) | Thermal Cyclers |
| Show More |