Learn More
Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatula
Product Code. 11347324
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
Product Code. 11347324
Brand: Schneider Gerd™ 251
Only null left
Specifications
Product Type Micro Spoon Spatula
Width (Metric) Blade 6 mm
Material Stainless Steel (4301)
Length (Metric) 130 mm