Learn More
Schneider Gerd™ 4301 Stainless Steel Micro Spoon Spatula
Product Code. 11387324
Click to view available options
Length (Metric):
210 mm
Unit Size:
Each
Product Code. 11387324
Brand: Schneider Gerd™ 6274
Only null left
Specifications
Product Type Micro Spoon Spatula
Width (Metric) Blade 7 mm
Dimensions (L x W) 210 x 7/7 x 10 mm
Material Stainless Steel (4301)
Length (Metric) 210 mm