Learn More
Schneider Gerd™ 4301 Stainless Steel Double Ended Spoon
Click to view available options
Length (Metric):
120 mm
180 mm
Unit Size:
Each
Description
- Spoons collect and transport dry substances or liquids
- Two end shapes offer increased versatility
- Ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
Specifications
Specifications
| Quantity | 1 Pk. |
| Product Type | Double Spoon End Spatula |
| For Use With (Application) | For Chemical |
| Material | Stainless Steel (4301) |
| Length (Metric) | 120 mm |