Learn More
Schneider Gerd™ 4301 Stainless Steel Double Ended Micro Spatula
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Description
- Stainless steel micro spatula
- Ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
Specifications
Specifications
| Quantity | 1 Pk. |
| Product Type | Double Ended Micro Spatula |
| Width (Metric) Blade | 5 mm |
| Dimensions (L x W) | 5 x 45 mm |
| Material | Stainless Steel (4301) |
| Length (Metric) | 150 mm |