missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 1000 μg/g in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
100 g
200 g
400 g
Unit Size:
100g
200g
400g
50g
Description
Metal Additives Standard, MA5: Ba, Ca, Mg, P, Zn @ 1000 μg/g in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Barium/Calcium/Magnesium/Phosphorus/Zinc |
| CAS | 7440-39-3, 7440-70-2, 7439-95-4, 7723-14-0, 7440-66-6 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Barium/Calcium/Magnesium/Phosphorus/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 75 cSt Hydrocarbon Oil |
| Quantity | 400 g |
| Concentration or Composition (by Analyte or Components) | Barium 1000 μg/g/Calcium 1000 μg/g/Magnesium 1000 μg/g/Phosphorus 1000 μg/g/Zinc 1000 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction