missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 40 kDa Phenyldextran, a Hydrophobic Dextran for Protein Interaction and Chromatography

Product Code. 30216516
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30216516 10 g 10g
30216364 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30216516 Supplier TdB Labs AB Supplier No. PhD40_10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phenyl dextran 40 kDa is a hydrophobic, aromatic-modified dextran used in protein interaction studies, chromatography, and surface modification. Its phenyl groups enable hydrophobic binding and phase separation in biochemical and analytical systems.

  • Product Overview: Phenyldextran 40 kDa is a hydrophobic dextran derivative designed for studies involving protein interactions, hydrophobic chromatography, and molecular binding research. The introduction of phenyl groups onto the dextran backbone provides hydrophobic character while maintaining water solubility, making it ideal for a range of biochemical and analytical applications.
  • Features:
    • Hydrophobic Modification: Phenyl groups impart hydrophobicity, enhancing interactions with nonpolar biomolecules and surfaces.
    • Molecular Weight: 40 kDa, balancing diffusion properties with sufficient binding surface area for interaction studies.
    • Water-Soluble: Despite its hydrophobic modification, Phenyldextran remains soluble in aqueous buffers, facilitating biological experiments.
    • Consistent Quality: Manufactured under controlled conditions to ensure reproducibility in protein binding and chromatography applications.
  • Benefits:
    • Ideal for Hydrophobic Interaction Studies: Models interactions between hydrophobic patches on proteins and hydrophobic surfaces.
    • Supports Chromatography Applications: Suitable for use in hydrophobic interaction chromatography (HIC) and related purification workflows.
    • Flexible Use: Can be applied in both analytical studies and preparative separations involving macromolecules.
    • Maintains Aqueous Compatibility: Offers the advantage of hydrophobic interaction while remaining compatible with biological systems.
  • Applications:
    • Hydrophobic interaction chromatography (HIC) for protein separation and purification.
    • Protein-protein and protein-surface interaction studies.
    • Modeling hydrophobic behavior of macromolecules in aqueous environments.
    • Development of specialized binding assays involving hydrophobic targets.
  • Technical Specifications:
    • Molecular Weight: Approximately 40 kDa.
    • Modification: Phenyl groups covalently attached to dextran backbone.
    • Appearance: Supplied as a dry, light-sensitive powder, fully water-soluble.
  • Storage and Stability:
    • Store dry at 2°C to 8°C, protected from light, to maintain optimal stability.
    • Stable for extended periods under recommended storage conditions.
  • Packaging and Availability:
    • Supplied as dry powder for easy handling and preparation.
    • Available in various research-grade packaging sizes.

Specifications

Quantity 10 g
Molecular Weight (g/mol) 40 kDa
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.