missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzoic acid, 98%

Product Code. p-7081220
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11468983

Brand: Thermo Scientific Alfa Aesar A11107.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1075-49-6 | C9H8O2 | 148.161 g/mol

It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in dichloromethane and methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-49-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00002569
InChI Key IRQWEODKXLDORP-UHFFFAOYSA-N
Synonym 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
PubChem CID 14098
IUPAC Name 4-ethenylbenzoic acid
SMILES C=CC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∼142°C
Density 1.3
Quantity 1 g
Beilstein 2041130
Sensitivity Light sensitive
Solubility Information Soluble in dichloromethane and methanol
Formula Weight 148.16
Percent Purity 98%
Chemical Name or Material 4-Vinylbenzoic acid
compliance-icons
Product Identifier
  • 4-Vinylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Vinylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.