missing translation for 'onlineSavingsMsg'
Learn More

4-Thiouridine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7041906
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
13957-31-8
C9H12N2O5S
260.26
MFCD00006538
ZLOIGESWDJYCTF-XVFCMESISA-N
3032615
CHEBI:20480
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O
This item is not returnable. View return policy
13957-31-8
C9H12N2O5S
260.26
MFCD00006538
ZLOIGESWDJYCTF-XVFCMESISA-N
3032615
CHEBI:20480
OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O

4-Thiouridine is a constituent of several transfer RNA's. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Thiouridine is a constituent of several transfer RNA′s. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

Solubility
Soluble in water (20 mg/ml), and methanol (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 25 mg
Synonym 4-thiouridine, thiouridine, uridine, 4-thio, 1-beta-d-ribofuranosyl-4-thiouracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, 1-beta-d-ribofuranosyl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, s4u, thiouridine van, uridine,4-thio, 4-thio-d-uridine
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
Formula Weight 260.27
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material 4-Thiouridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'