missing translation for 'onlineSavingsMsg'
Learn More

(4-((Pyrrolidin-1-ylsulfonyl)methyl)phenyl)carbamic Acid tert-Butyl Ester, TRC

Product Code. 30427446
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427446 100 mg 100mg
30417734 25 mg 25mg
30416184 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427446 Supplier Toronto Research Chemicals Supplier No. TRCP997935100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-((Pyrrolidin-1-ylsulfonyl)methyl)phenyl)carbamic Acid tert-Butyl Ester
Analyte or Component Names (4-((Pyrrolidin-1-ylsulfonyl)methyl)phenyl)carbamic Acid tert-Butyl Ester
Shipping Condition Room Temperature
Molecular Formula C16H24N2O4S
Solution Type Neat
Industry Type Pharmaceutical
Molecular Weight (g/mol) 340.44
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.