missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidineethanol, 97%

Product Code. p-2716131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307241 1 g Glass bottle 1g
10355851 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307241 Supplier Thermo Scientific Supplier No. 263650010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-26-4 | C7H16NO | 130.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-26-4
Molecular Formula C7H16NO
Molecular Weight (g/mol) 130.21
MDL Number MFCD00006008
InChI Key LDSQQXKSEFZAPE-UHFFFAOYSA-O
Synonym 4-piperidineethanol, 2-piperidin-4-yl ethanol, 4-2-hydroxyethyl piperidine, 2-piperidin-4-yl ethan-1-ol, 4-piperidine ethanol, piperidine-4-ethanol, 4-ethanolpiperidine, 2-4-piperidyl ethanol, 2-piperidin-4-yl-ethanol, 4-hydroxyethyl-piperidine
PubChem CID 73953
SMILES OCCC1CC[NH2+]CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Boiling Point 131.0°C to 136.0°C (17.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4902
Quantity 1 g
Beilstein 21, 4
IUPAC Name 2-piperidin-4-ylethanol
Formula Weight 129.19
Percent Purity 97%
Chemical Name or Material 4-Piperidineethanol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Piperidineethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.