missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinnamaldehyde, predominantly trans, 98%

Product Code. 11364689
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
This item is not returnable. View return policy

Product Code. 11364689

Brand: Thermo Scientific Alfa Aesar A11467.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1734-79-8 | C9H7NO3 | 177.159 g/mol

Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4'-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2?-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4′-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2′-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1734-79-8
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00007379
InChI Key ALGQVMMYDWQDEC-OWOJBTEDSA-N
Synonym 4-nitrocinnamaldehyde, 3-4-nitrophenyl acrylaldehyde, p-nitrocinnamaldehyde, 2-propenal, 3-4-nitrophenyl, cinnamaldehyde, p-nitro, trans-4-nitrocinnamaldehyde, e-3-4-nitrophenyl acrylaldehyde, 3-4-nitrophenyl-2-propenal, unii-48r8lam7yx, 2e-3-4-nitrophenyl prop-2-enal
PubChem CID 5354135
IUPAC Name (E)-3-(4-nitrophenyl)prop-2-enal
SMILES C1=CC(=CC=C1C=CC=O)[N+](=O)[O-]

Specifications

Melting Point 138°C to 142°C
Odor Characteristic
Quantity 50 g
Beilstein 1565424
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 4-Nitrocinnamaldehyde, predominantly trans

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.