missing translation for 'onlineSavingsMsg'
Learn More

4-n-Heptyloxybenzoic acid, 98%, Thermo Scientific™

Product Code. 11384689
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384689 10 g 10g
11467976 25 g 25g
11477976 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384689 Supplier Thermo Scientific Alfa Aesar Supplier No. A14059.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15872-42-1
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.31
MDL Number MFCD00013992
InChI Key ZRVIYEJYXIDATJ-UHFFFAOYSA-N
Synonym 4-heptyloxy benzoic acid, 4-heptyloxybenzoic acid, 4-n-heptyloxybenzoic acid, p-heptoxybenzoic acid, p-heptyloxybenzoic acid, benzoic acid, 4-heptyloxy, unii-bj0yzc8zy8, benzoic acid, p-heptyloxy, p-n-heptyloxybenzoic acid, bj0yzc8zy8
PubChem CID 85154
ChEBI CHEBI:34405
IUPAC Name 4-heptoxybenzoic acid
SMILES CCCCCCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 92°C to 94°C
Quantity 10 g
Beilstein 2211867
Formula Weight 236.31
Percent Purity 98%
Chemical Name or Material 4-n-Heptyloxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.