Learn More
4-Methoxypyridazin-3-amine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxypyridazin-3-amine |
| CAS | 1026194-47-7 |
| Molecular Formula | C5 H7 N3 O |
| Quantity | 50 mg |
| Synonym | 3-Pyridazinamine, 4-methoxy- (ACI), 4-Methoxy-3-pyridazinamine (ACI) |
| InChI Formula | InChI=1S/C5H7N3O/c1-9-4-2-3-7-8-5(4)6/h2-3H,1H3,(H2,6,8) |
| SMILES | COc1ccnnc1N |
| IUPAC Name | 4-methoxypyridazin-3-amine |
| Molecular Weight (g/mol) | 125.128 |
| Formula Weight | 125.0589 |