missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-(1-naphthyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15492916
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15492916 250 mg 250mg
15403368 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15492916

Brand: Thermo Scientific Alfa Aesar H56822.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(1-Naphthyl)benzenesulfonamides efficiently couple with internal alkynes via cleavage of the C-H bond at the peri-position in the presence of a catalyst system of [IrCl(cod)]2/PBut3 to selectively afford the corresponding 8-vinyl-1-naphthol derivatives.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56799-97-4
Molecular Formula C17H15NO3S
Molecular Weight (g/mol) 313.371
MDL Number MFCD00682767
InChI Key KBIAXNFZLJUXNY-UHFFFAOYSA-N
Synonym 4-methoxy-n-1-naphthyl benzenesulfonamide, 4-methoxy-n-naphthalen-1-yl benzenesulfonamide
PubChem CID 899644
IUPAC Name 4-methoxy-N-naphthalen-1-ylbenzenesulfonamide
SMILES COC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC3=CC=CC=C32

Specifications

Quantity 250 mg
Beilstein 3036729
Formula Weight 313.37
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-(1-naphthyl)benzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.