Learn More
4-Isopropylaniline, 99%
Product Code. p-7022052
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11427606
Brand: Thermo Scientific Alfa Aesar A14646.14
Request Bulk or Custom Format
CAS: 99-88-7 | C9H13N | 135.21 g/mol
4-Isopropylaniline was used in the hydroamination of phenylacetylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Isopropylaniline was used in the hydroamination of phenylacetylene.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong acids. Light sensitive
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99-88-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007900
InChI Key LRTFPLFDLJYEKT-UHFFFAOYSA-N
Synonym 4-isopropylaniline, cumidine, 4-aminocumene, p-isopropylaniline, p-cumidine, 4-propan-2-yl aniline, para-isopropylaniline, 4-amino-1-isopropylbenzene, cumene, p-amino, aniline, p-isopropyl
PubChem CID 7464
ChEBI CHEBI:43405
IUPAC Name 4-propan-2-ylaniline
SMILES CC(C)C1=CC=C(C=C1)N
Specifications
Melting Point -63°C
Density 0.989
Boiling Point 226°C to 228°C
Flash Point 92°C (197°F)
Refractive Index 1.543
Quantity 25 g
Beilstein 1930544
Sensitivity Light sensitive
Merck Index 14,2620
Solubility Information Insoluble in water.
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material 4-Isopropylaniline