Learn More
4-Hydroxybenzamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzamide |
| CAS | 619-57-8 |
| Molecular Formula | C7H7NO2 |
| Quantity | 25 g |
| Synonym | 4-Carbamoylphenol, NSC 524134, SR 4328, p-Hydroxybenzamide, p-Hydroxybenzoic Acid Amide |
| InChI Formula | InChI=1S/C7H7NO2/c8-7(10)5-1-3-6(9)4-2-5/h1-4,9H,(H2,8,10) |
| SMILES | NC(=O)c1ccc(O)cc1 |
| IUPAC Name | 4-hydroxybenzamide |
| Molecular Weight (g/mol) | 137.14 |
| Formula Weight | 137.0477 |