Learn More
4-Hydroxy Valproic Acid Sodium Salt(Mixture of Diastereomers), TRC
Product Code. 30180403
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180403 1 g 1g
30180260 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30180403 Supplier Toronto Research Chemicals Supplier No. TRCH9930051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1216888-06-0
Molecular Formula C8 H15 O3 . Na
Molecular Weight (g/mol) 182.19
Synonym 4-Hydroxy-2-propyl-pentanoic Acid Sodium Salt, 2-Propyl-4-hydroxypentanoic Acid Sodium Salt, 2-n-Propyl-4-hydroxypentanoic Acid Sodium Salt
IUPAC Name sodium;4-hydroxy-2-propylpentanoate
SMILES [Na+].CCCC(CC(C)O)C(=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H16O3.Na/c1-3-4-7(8(10)11)5-6(2)9;/h6-7,9H,3-5H2,1-2H3,(H,10,11);/q;+1/p-1
Formula Weight 182.0919
Chemical Name or Material 4-Hydroxy Valproic Acid Sodium Salt (mixture of diastereomers)