missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl Carbamic Acid Molindone Ester, TRC

Product Code. 30407986
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407986 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407986 Supplier Toronto Research Chemicals Supplier No. TRCF632083250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Fluorophenyl Carbamic Acid Molindone Ester
Analyte or Component Names 4-Fluorophenyl Carbamic Acid Molindone Ester
Shipping Condition Room Temperature
Molecular Formula C23 H27 F N2 O4
InChI Formula InChI=1S/C23H27FN2O4/c1-3-19-15(2)26(23(28)30-18-7-5-17(24)6-8-18)20-9-4-16(22(27)21(19)20)14-25-10-12-29-13-11-25/h5-8,16H,3-4,9-14H2,1-2H3
Solution Type Neat
SMILES CCc1c(C)n(C(=O)Oc2ccc(F)cc2)c3CCC(CN4CCOCC4)C(=O)c13
IUPAC Name (4-fluorophenyl) 3-ethyl-2-methyl-5-(morpholin-4-ylmethyl)-4-oxo-6,7-dihydro-5H-indole-1-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 414.47
Quantity 250 mg
Formula Weight 414.195
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.