Learn More
4-Dimethylaminobutyraldehyde Diethyl Acetal, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Dimethylaminobutyraldehyde Diethyl Acetal |
| CAS | 1116-77-4 |
| Molecular Formula | C11H23NO2 |
| Quantity | 100 g |
| Synonym | 4,4-Diethoxy-N,N-dimethyl-1-butanamine, 1-(N,N-Dimethylamino)-4,4-diethoxybutane, 4,4-Diethoxy-N,N-dimethylbutanamine, 4,4-Diethoxy-N,N-dimethylbutylamine, 4-(Dimethylamino)butanal Diethyl Acetal, Zolmitriptan USP Related Compound H |
| InChI Formula | InChI=1S/C10H23NO2/c1-5-12-10(13-6-2)8-7-9-11(3)4/h10H,5-9H2,1-4H3 |
| SMILES | CCOC(CCCN(C)C)OCC |
| Molecular Weight (g/mol) | 189.3 |
| Formula Weight | 189.1729 |