missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobenzaldoxime, 97%, Thermo Scientific™

Product Code. 11332127
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11332127

Brand: Thermo Scientific Alfa Aesar L07961.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54376-65-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00051801
InChI Key DHNWBEYWLRLFHM-FMIVXFBMSA-N
Synonym 4-diethylaminobenzaldoxime, 4-diethylamino benzaldehyde oxime, n,n-diethyl-4-1e-hydroxyimino methyl aniline, 4-diethylaminobenzaldehyde oxime, n,n-diethyl-4-hydroxyimino methyl aniline, 4-diethylaminobenzaldehydeoxime, diethyl 4-hydroxyimino methyl phenyl amine, n,n-diethyl-4-e-hydroxyimino methyl aniline, n,n-diethyl-4-1z-hydroxyimino methyl aniline, ne-n-4-diethylamino phenyl methylidene hydroxylamine
PubChem CID 6876690
IUPAC Name (NE)-N-[[4-(diethylamino)phenyl]methylidene]hydroxylamine
SMILES CCN(CC)C1=CC=C(C=C1)C=NO

Specifications

Melting Point 94°C to 97°C
Quantity 5 g
Beilstein 2722071
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 4-Diethylaminobenzaldoxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.