Learn More
(4-Cyanobenzyl)triphenylphosphonium chloride, 99%
Product Code. 11388655
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388655 5 g 5g
11398655 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11388655 Supplier Thermo Scientific Alfa Aesar Supplier No. L12187.06
Request Bulk or Custom Format
CAS: 20430-33-5 | C26H21ClNP | 413.88 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20430-33-5
Molecular Formula C26H21ClNP
Molecular Weight (g/mol) 413.88
MDL Number MFCD00011752
InChI Key GXMFPJIKPZDNCS-UHFFFAOYSA-M
Synonym 4-cyanobenzyl triphenylphosphonium chloride, 4-cyanophenyl methyl triphenylphosphanium chloride, 4-triphenylphosphino methyl benzenecarbonitrile, chloride, acmc-20apna, phosphonium, 4-cyanophenyl methyl triphenyl-, chloride, 4-cyanobenzyltriphenylphosphonium chloride, p-cyanobenzyl-triphenylphosphonium chloride, 4-cyanobenzyl triphenylphosphoniumchloride, 4-cyanobenzyl-triphenylphosphonium chloride
PubChem CID 2724468
SMILES [Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Specifications
Melting Point 322°C to 326°C
Odor Odorless
Quantity 5 g
Beilstein 4630137
IUPAC Name (4-cyanophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 413.89
Percent Purity 99%
Chemical Name or Material (4-Cyanobenzyl)triphenylphosphonium chloride